Learn More
MilliporeSigma™ Chemicon™ Caspase 3 Colorimetric Assay Kit, DEVD
Provide a simple and convenient means for assaying the activity of caspases that recognize the sequence DEVD
Supplier: MilliporeSigma™ APT165
Specifications
-20°C | |
Chromogenic | |
pNA Standard, 5X Cell Lysis Buffer, 5X Assay Buffer, Caspase-3 Substrate, Caspase-3 Inhibitor |
Caspase 3 Colorimetric Assay Kit, DEVD | |
Provide a simple and convenient means for assaying the activity of caspases that recognize the sequence DEVD | |
100 Tests |