Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ Reagents for Electrophoresis: Melt and Pour UltraSpec-Agarose™
Complements the majority of EDVO-Kits that involve DNA electrophoresis - Edvotek™ Reagents for Electrophoresis: Melt and Pour UltraSpec-Agarose™ is sufficient for the separation of DNA fragments in the size range of 500-30,000 base pairs.
$38.75 - $191.00
Specifications
Buffer | TAE |
---|---|
Concentration | 0.8% |
Format | Melt and Pour |
Product Type | Agarose |
For Use With (Application) | Agarose or Polyacrylamide Gel electrophoresis |
Description
- Melt and Pour UltraSpec-Agarose™ consists of 0.8% electrophoretic grade UltraSpec-Agarose™ in tris-acetate-EDTA buffer
Specifications
TAE | |
Melt and Pour | |
Agarose or Polyacrylamide Gel electrophoresis |
0.8% | |
Agarose | |
Agarose |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title