Learn More
DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide cured silicone
Catalog Number 02900141
Click to view available options
Thread Size (Metric):
GL 18
GL 25
GL 32
Cap liners with PTFE faced silicone.
Replacement cap liners suitable for PBT screw caps and tamper-evident screw caps. Heat resistance to 130°C (steam) and 200°C (dry heat). Excellent chemical resistance due to PTFE coating. Silicon is peroxide catalysed.
  • replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
  • Excellent chemical resistance due to PTFE coating
  • Cap liner forms a tight seal with bottle Peroxide catalysed silicone
  • Peroxide catalysed silicone
  • Suitable for PBT screw caps and tamper-evident screw caps
Diameter (Metric) 30.5 mm
For Use With (Application) Replacement cap liner for red PBT caps
Material Silicone, PTFE, VMQ
Quantity 10/Pk.
Thread Size (Metric) GL 32
Autoclavable Autoclavable