Learn More
Siglent Technologies DUAL-CHANNEL DIG OSCILLOSCOPE
Supplier:  Siglent Technologies SDS1052DL+
Encompass_Preferred
SDS1000DL+/CML+ series is a dual-channel universal digital oscilloscope, available in 50 MHz, 100 MHz and 150 MHz bandwidth models. It includes a 2 Mpts memory depth that helps to ensure accurate waveform resolution and to capture longer signal lengths. With its 7 inch TFT-LCD (800*480) screen, there is adequate screen space to help better see and analyze waveform details. Along with a 1 GSa/s sampling rate, the SDS1000CML+ supports 32 parameters measurements and common mathematical operations to speed up complex / repetitive measurements.
Catalog No. NC1602330