Learn More
Drummond™ Repeat Kit with Valve
Supplier: Drummond™ 3000091
Specifications
Microvalve, 3 way valve, Stainless Steel sinker, 30 in. Tygon tubing, luer pipet tips, 7 Polypropylene tubing connectors | |
Pipetting Repeat Kit | |
Models 390 and 590 Drummond™ microdispensers |