Learn More
United States Biological Corporation Drop-out Base with Glucose and Agar, Powder, 100 g
Supplier: United States Biological Corporation D951001100G
Nitrogen, carbon, trace element, and vitamin source for yeast growth used as a media base for yeast transformant selection. Supplement with the appropriate DO Mix for selection and propagation of yeast transformants. Glucose allows for the rapid growth of yeast with short doubling times.