Learn More
Cole-Parmer Disposable BOD Bottle, 300 ml, 100/pk
Supplier:  Cole-Parmer 5320002
Encompass
Environmental Express Disposable BOD Bottle, 300 ml, 100/pk, Get more accurate results and never wash a BOD bottle againLaboratory tested and EPA method 5210 B approved BOD methodIndependent study confirms improved blanks, improved results, and improved precisionReduces costs for labor and for water consumptionEasily identify samples with individually numbered bottles
Catalog No. NC1226630