Learn More
Bachem H-Asp-Asp-OH
SDP
Supplier:  Bachem 4010210.1000
Encompass_Preferred
H-Asp-Asp-OH, 58471-53-7, C8H12N2O7, Mr 248.19, 1g. Dipeptides as aspartylaspartate and glutamylglutamate can be utilized as growth substrates for the oral bacteria Porphyromonas gingivalis, Prevotella intermedia, Prevotella nigrescens and Fusobacterium nucleatum.
Catalog No. 50-194-6825