Learn More
Anton Paar Measuring system adapter for ViscoQC 100 & ViscoQC 300
Supplier: Anton Paar 191719
Adapter required for measurement with a concentric cylinder (CC), double-gap (DG) or SSA (SC4) measuring system if not using PTD 80.Use with Measuring System CC12 item NC1818606, CC18 item NC1818604, CC26 item NC1818602, CC12/D18 item NC1818608, SC4-18 item NC1818611, SC4-21 item NC1818613, SC4-27 item NC1818615, DG26 item NC1818619, or DG26/SS NC1818621Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.