Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Digital Meters
SCI_ED

Designed for simple, accurate test readings.

Specifications

For Use With (Application) pH
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S02336
SCI_ED
View Documents
Luster Leaf Products, Inc.
1825
Printed instructions and extensive plant list packed with the meter Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
S02337
SCI_ED
View Documents
Luster Leaf Products, Inc.
1845
Printed instructions and plant pH preference list packed with the meter Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Digital Moisture Meter

  • New digital results from 0 to 9.9
  • Printed instructions and extensive plant list packed with the meter

Digital pH Meter
  • New digital results from 3.5 to 9.0
  • Printed instructions and plant pH preference list packed with the meter

Digital PLUS Moisture Meter
  • New digital results from 0 to 9.9
  • Extensive plant list with preferred soil moisture levels and special plant needs built in to meter database

Digital PLUS Soil pH Meter
  • New digital results from 3.5 to 9.0
  • Plant pH preference list is built in to the meter database

Note: Probe should not be left in contact with moisture for extended periods of time.

Specifications

Specifications

pH
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit