Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Small Round, 1.3 mm Tip, 013 I.S.O.
Supplier:  Electron Microscopy Sciences 6215513
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 1.3 mm
I.S.O. size: 013
Catalog No. 50-254-40