Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ DF300 Flat-Fold N95 Respirator, Headstrap
Engineered to promote easier breathing, the DF300 Series Flatfold Disposable Respirators are made with a multi-layered absorption media that repels moisture and features a smooth inner lining for added comfort.
Supplier: Honeywell™ DF300H910N95
This item is not returnable.
View return policy
Description
- Designed to fit all faces comfortably: and accommodate the user's facial movements
- Multi-layered absorption media with a smooth inner lining for added comfort
- Latex free woven head straps are welded for a secure fit
- Concealed: adjustable nose clip and soft foam nose cushion provide a custom secure seal
- Humidity and moisture resistant filter media that repels moisture for a longer lasting respirator
- Reliable: durable design that lasts throughout the workday
Specifications
Flat-fold | |
N95 | |
General Purpose Particulate Respirator | |
No |
Single-use | |
Multi-Layered Absorption Media | |
One Size | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction