Learn More
Thermo Scientific Cuvettes 10PK
Supplier:  Thermo Scientific 986540TS
Encompass
Disposable rectangular plastic sample cells with clear optical windows for spectroscopic analysis of liquid samples, 10MM light path, made of optical grade polystyrene for UV transparency down to 340NM, 3.5ML volume capacity, frosted sides, easy handling and cleaning, used for fluorescence or luminescence measurements.
Catalog No. NC2227376