Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Smart Lock Connected Leading Edge Self-Retracting Lifeline

The first digitally connected SRL in the fall protection industry

$1694.00 - $2370.00

Specifications

Description The 3M™ DBI-SALA™ Smart Lock Connected Self-Retracting Lifeline is designed to help increase your safety intelligence with integrated connected capability for smart PPE management. You can now know the date and time of brake events. It also tracks usage—how many extensions and, pawl locks, and how much active time, the device has experienced, and when—a fall protection industry first.
Color Black
Material Galvanized Steel
Product Type Leading Edge Self-Retracting Lifeline- Cable
For Use With (Application) Leading Edge, Decking
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
19-070-977
View Documents
3M™
3503843
20 ft.
Case for $1,694.00
Add to Cart
 
19-070-976
View Documents
3M™
3503842
30 ft.
Case for $2,370.00
Add to Cart
 
Description

Description

  • Brake event notifications sent via the mobile/web app, and a service light on the SRL, help improve incident reporting and response
  • Time stamped usage data (extensions, pawl locks, brake events) can help confirm when and how the SRL has been used
  • Get equipment status and usage statistics from multiple SRLs to enhance inventory management
  • Patented magnetic retraction control system allows lifeline to retract smoothly and hands free
  • Patented dual label system located within ergonomic cable handle simplifies inspections and reduces the need for replacement
  • Using advanced motion capture, horizontal use is designed to reduce nuisance lockups when compared to previous generation SRLs
  • Rugged and extremely lightweight thermoplastic housing with ergonomic handle
Specifications

Specifications

The 3M™ DBI-SALA™ Smart Lock Connected Self-Retracting Lifeline is designed to help increase your safety intelligence with integrated connected capability for smart PPE management. You can now know the date and time of brake events. It also tracks usage—how many extensions and, pawl locks, and how much active time, the device has experienced, and when—a fall protection industry first.
Galvanized Steel
Leading Edge, Decking
420 lb.
Black
Leading Edge Self-Retracting Lifeline- Cable
Carabiner
190 kg
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.