Learn More
3M™ DBI-Sala™ Delta™ Vest Style Harnesses
No-Tangle™ triangular design creates frame that allows the harness to fall into place when donned
Supplier: 3M™ 70007412201
Description
- Spring-loaded stand-up dorsal D-ring design ensures fast, easy and safe connections
- Rip-stitch indicators allow to inspect the harness at glance, for prior damaging impact loads
- Harness falls into place, allowing quick and easy donning
- Side D-rings for positioning
- Adjustable non-slip chest strap with easy to use pass-thru buckle
- Tongue buckle leg straps for ease of use and will not loose
- Lanyard keepers holds unused snap hooks and help reduce trip hazard
- Revolver vertical torso adjuster provides simple and fast adjustment eliminating loose ends and locks webbing into place preventing slippage
- Velcro cover protects harness labels and holds I-Safe™ ID tag
Compliance
Meets ANSI™ Z359.11 requirements.
Specifications
Pass-Through | |
Tongue Buckle | |
Vest Style |
Back | |
Large |