Learn More
Bachem (D-Arg1,D-Trp7·9,Leu11)-Substance P
SDP
Supplier:  Bachem 4004691.0001
Encompass_Preferred
(D-Arg1,D-Trp7·9,Leu11)-Substance P, H-D-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2, 91224-37-2, C75H108N20O13, Mr 1497.81, 1mg. Spantide I, [DArg1, DTrp7,9, Leu11] Substance P. Spantide I, a high potency substance P antagonist with a weak spasmogenic and a poor histamine releasing effect.
Catalog No. 50-259-551