Learn More
Perkin Elmer US LLC Special Optical Glass Cylindrical Cell with PTFE Stopper - 50 mm, Qty. 1
Supplier:  Perkin Elmer US LLC B0631093
Encompass_Preferred
Cylindrical cells have an outside diameter of 22 mm and an inside diameter of 19 mm. A package contains one cell and two windows. Special Optical Glass cells are made from exceptionally pure raw materials and have a spectral range of 320 - 2,500 nm. These cells are identified by a green PerkinElmer logo.
Catalog No. 50-904-2651