Learn More
INDUSTRIAL TEST SYSTEMS INC Cyanide Reagent Test Strip Kit
SDP
Supplier:  INDUSTRIAL TEST SYSTEMS INC 484003
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Kit: ReagentStrip #1 (bottle of 50), ReagentStrip #2 (bottle of 50),4 cuvettes, pipette, CD, instructions/MSDS, and 2 color charts.
Catalog No. NC9251715