Learn More
Fireflysci Type P65S (Single Cuvette Washer)
SDP
Supplier:  Fireflysci P65S
Encompass_Preferred
Cuvette washer head for power washing a cuvette• Designed to quickly clean and dry cuvettes with no handling involved• Comes with gasket and stopper 7• Can clean cuvettes from 3mm inner diameter up to 10mm• Vacuum flask, vinyl tubing and pump sold separately
Catalog No. 50-106-0453