Learn More
R&D Systems™ Cultrex Laminin I Cell Invasion Assay, 24-well
Asseses cell invasion through Laminin I.
Supplier: R&D Systems™ 3456024K
This item is not returnable.
View return policy
Description
Standardized assay for Laminin I cell invasionAvailable in 24 and 96 well formatsAllows optimization for specific cell linesSpecifications
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
24 Tests |