Learn More
CORNING SPECIALTY MATERIALS Corning microHSI™ 410 Vis-NIR Hyperspectral Sensor
Supplier:  CORNING SPECIALTY MATERIALS 11971100
Encompass_Preferred
Request Bulk Order
Hyperspectral sensor; The Corning microHSI™ 410 Vis-NIR hyperspectral sensor is a push-broom line imaging spectrometer designed for the 400nm-1000nm spectral range. This system has a higher optical numerical aperture (NA) for enhanced light collection sensitivity in an extremely compact, lightweight package. Cornings patented hyperspectral system delivers stability over temperature, pressure, and humidity with significantly better throughput, superior sensitivity to light, and durable packaging for consistent, reliable results. This is a solid block offner spectrograph with diamond-ruled high-efficiency reflective blazed grating with a 29.5 degree standard full field of view. The microHSI™ 410 Vis-NIR hyperspectral sensor is ideal for applications including process control, precision agriculture, medical diagnostics, food quality, and more. Dimensions are 15.4x2.5x2.4 with standard foreoptic and 4.0x2.5x2.4 without foreoptic and weight is 1 lb.
Catalog No. 50-152-8531
Please call customer service for current pricing. \n1-800-766-7000