Learn More
Corning™ Costar™ Transtar™-96 96-Well Liquid Transfer System Accessories: Reservoir Liners
GREENER_CHOICE
For use with 96-well plates
$274.00 - $292.00
Specifications
Sterility Sterile
For Use With (Application) Ideal for repetitively filling multi-channel pipettors
No. per Case 100/Cs.
No. per Pack 1/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
07-200-56
GREENER_CHOICE
View Documents
Corning™
4876
Open Reservoir Liner
Case of 100 for $292.00
Add to Cart
07-200-58
GREENER_CHOICE
View Documents
Corning™
4878
8 Channel Reservoir
Case of 100 for $285.00
Add to Cart
07-200-57
View Documents
Corning™
4877
12 Channel Reservoir
Case of 100 for $274.00
Add to Cart
Description
Description
For use with 96-well plates
These are sterile replacement open reservoir liners for the Transtar-96™ Well Liquid Transfer system.
Specifications
Specifications
Sterile
Ideal for repetitively filling multi-channel pipettors
100/Cs.
1/Pk.