Learn More
Electron Microscopy Sciences Embra Coordinator 200 mesh Grid, Copper
Supplier:  Electron Microscopy Sciences EC200CU
Encompass_Preferred
Standard 3.05 mm diameter grids with a handle.
Grid: Copper
Catalog No. 50-251-18