Learn More
Santa Cruz Biotechnology CONTROL SIRNA-A
Supplier:  Santa Cruz Biotechnology SC37007
Encompass
Control siRNA-A, resuspend to 66uL, 10uM, Sufficient for 10 to 20 transfections (in 12 well plate), suitable as a negative control for experiments using targeted siRNA transfection
Catalog No. NC9175272