Learn More
Chemglass Life Sciences 50L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 16.2" Clearance Below Valve
SDP
Supplier:  Chemglass Life Sciences CG1968650
Encompass_Preferred
Request Bulk Order
50L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 16.2" Clearance Below Valve. Features:• Motor Mount Arm Can Unlock & Swing Out Giving Greater Access To Reactor• Digital Temperature Monitor• Electric And XP Motor Has Separate Controller W/ Digital Display• Smaller Footprint Support Frame With Oversized Rugged S.S.Uprights And Heavy Duty PTFE Coated Aluminum Fittings• Reactor Jacket Graduation Scale On Both Sides• Lower Safety Ring• Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Arrows For Open & Close• Electric Or Air Motor Optional XP Motor Available• Open Frame Design Has Excellent Accessibility• 7-Neck Lid With Large Powder Addition Port• Support Plate Holds Reactor Securely & Allows Easy Installation/Removal.• Ridge Clamp Accepts 1/2” Rod And Adds Additional Support For Large Glassware• S.S.Inlet/Outlet Adapters Available In M16 X 1 And M30 X 1.5 Allow Connection To Cooling/Heating Circulator• Heavy Duty Lockable Casters
Catalog No. CG1968650