Learn More
Teledyne ISCO, Inc COLUMN GOLD C18 900 G
Supplier: Teledyne ISCO, Inc 692203492
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
950 gram, RediSep Rf Gold C18, 30 um columns, pkg. of 1