Learn More
Caron Products And Services CO2 Incubator, Shaker Ready, 25 cu. ft., Temp, No RH
Supplier:  Caron Products And Services 7401252
Encompass_Preferred
Request Bulk Order
Carons 2-Tier, Shaker-Ready CO2 Incubator is optimized for large-scale suspension cell culture applications using shakers. Standard features: 2 shaker-support shelves, internal outlets, intuitive touchscreen controls, IR CO2 sensor, and wide temp range (20-60C). 708L (25 ft3); 230V, 60Hz.
Catalog No. NC2676807