Learn More
Tritech Research Inc Drosophila Anesthesia CO2 Blow gun
Supplier:  Tritech Research Inc MINJDROSGUN
Encompass
Drosophila Anesthesia CO2 Blow gunThe pistol grip blowgun offers variable flow control and is lightweight, ergonomic and very intuitive to use. The simple trigger allows the user to deliver just the right amount of carbon dioxide to keep flies asleep. The convenient needle tip is designed to be slid into fly vials past cotton plugs (cotton, rayon, or Flugs) or caps for easy anesthetization. The blowgun will save your lab on CO2 costs compared with continuous-flow systems, and is much less likely to inadvertently kill important specimens due to over-anaesthetizing. It is safer than using organic solvents as anaesthetics.
Catalog No. NC1710679