Learn More
STEMCELL Technologies ClonaCell™-HY Medium E, Size: 500 mL
Supplier: STEMCELL Technologies 03805
ClonaCell™-HY Medium E is a serum-containing, nutritionally rich liquid medium optimized for hybridoma expansion after hypoxanthine, aminopterin, thymidine (HAT) selection. The medium contains hypoxanthine and thymidine (HT) and is used to wean hybridomas off aminopterin used during the selection process. This medium has been verified for use in mouse and rat hybridoma development and monoclonal antibody production and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster. ClonaCell™-HY Medium E has proven effective for subcloning and stabilization of unstable hybridoma cell lines.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.