Learn More
Gilson Company Inc Cleveland Flash Tester (115V / 60Hz)
Supplier:  Gilson Company Inc PT6A
Encompass
Cleveland Flash Point Tester is used in the Cleveland Open Cup method to measure flash and fire points for petroleum products igniting above 175°F (79°C).
Catalog No. NC1597997