Learn More
Electron Microscopy Sciences Micro Cleveland Bone Rongeur, 5 mm
Supplier: Electron Microscopy Sciences 7798605
Bone Rongeur with curved jaws. Made from stainless steel with a polished finish. Curved jaws, with a 5mm jaw width, and 7" long.