Learn More
United States Biological Corporation Chick Embryo Extract, Ultrafiltrate, Liquid, 20 mL
SDP
Supplier:  United States Biological Corporation C399920ML
Encompass_Preferred
Chick Embryo Extract, Ultrafiltrate is used as a supplement in some growth media formulations. It is prepared by blending 11-14 day old chick embryos in a balanced salt solution (3X embryo volume). The solution undergoes centrifugation to remove larger particles and debris. The supernatant is subjected to an ultrafiltration step with a 10kD MW cutoff to remove protein from the solution, producing a clear, amber liquid. This liquid is sterile-filtered.
Catalog No. NC1202490