Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scott Safety™ Charging Systems for Pak-Tracker Firefighting Locator System
Truck charging system chargers or recharging the nickel-metal hydride battery pack
$768.88 - $867.73
Catalog Number | Mfr. No. | Product Type | Voltage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Voltage | Price | Quantity & Availability | ||||
19-824-455
|
Scott Safety™
7012471473 |
Truck Charging System | 12 V |
Each for $867.73
|
|
||||
19-824-456
|
Scott Safety™
7012471475 |
Desktop Charging System | 110 V |
Each for $768.88
|
|
||||
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title