Learn More
Santa Cruz Biotechnology Double End, Flat Blade & Narrow Blade, Sterile | 100/case
Supplier:  Santa Cruz Biotechnology SC395251
Encompass
Cell Scrapers and Lifters are designed for harvesting cells from tissue culture flasks, dishes, plates or bottles. Cell Scrapers and Cell Lifters are non-pyrogenic and packaged sterile (gamma irradiated) in individual paper/poly wrappers with 100 pieces per case. Double Ended Cell Lifters have a 19mm beveled edge blade on one end and a J-Hook or Narrow Blade on the other for removing cells from Multiple Well Plates or Tubes
Catalog No. NC1870622