Learn More
Immunology Consultants Cat IgG Enriched, 20.0 mg
Supplier:  Immunology Consultants RA20G
Encompass_Preferred
The Ig fraction was enriched from normal cat serum using a combination of procedures that include salt fractionation. Antibody concentration was determined by extinction coefficient: O.D. 1.4 at 280nM equals 1.0 mg of IgG. Estimated purity of >80% as determined by SDSPAGE. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4320