Learn More
Carolina™ Carosafe™ Holding Solution
For use in formalin-preserved specimens. Carolina™ Carosafe™ Holding Solution is formaldehyde-free and cannot be used to fix specimens.
Supplier: Carolina™ 853341
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- Carosafe preservative and holding solution for biological specimens contains no formaldehyde and is not a tissue fixative
- This produces a formalin-preserved specimen that, when dissected, minimizes formaldehyde exposure
Order Info
For use in science education laboratories – no certificate of analysis is available; Ready-to-use; do not dilute; Health Risk: 1; Flammability: 1; Reactivity: 0
Specifications
Ready to use. | |
4 L | |
Preservative and holding solution for biological specimens |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.