Learn More
Cargille Consumables and Reagents for Albumin Test Set
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For use with albumin test kit
Supplier: Cargille 52711/1000
Description
- Specimen tubes are graduated at 4.5mL (0.15 oz., for distilled water and a sulfosalicylic acid tablet) and at 6mL (0.20 oz., for addition of 1.5mL [0.05 oz.] of urine) to eliminate the need for pre-measuring
Specifications
Salicylic Acid Tablet | |
Albumin test kit | |
1000 Tablets |