Learn More
C.L. Sturkey Gold Disposable Microtome Blades
The ultimate blade for cutting hard tissue and bone
$134.00 - $394.00
Specifications
Disposable Yes
Coating Titanium Nitride
View More Specs
Products 4
Catalog Number Mfr. No. Profile Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Profile Quantity Price Quantity & Availability  
22-210013
Accessories View Documents
C.L. Sturkey
D554G
High 100 Pk.
Pack of 100 for $276.00
Add to Cart
22-210009
Accessories View Documents
C.L. Sturkey
DT315G
Low 170 Pk.
Pack of 170 for $394.00
Add to Cart
22-210-049
View Documents
C.L. Sturkey
DT315G50
Low 50 Pk.
Pack of 50 for $151.00
Case of 10 PK for $1,344.00
Edge
Add to Cart
22-210-046
View Documents
C.L. Sturkey
D554G50
High 50 Pk.
Pack of 50 for $134.00
Case of 10 PK for $1,264.00
Add to Cart
Description
Description
High-performance specialty blade, tough and durable
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Specifications
Yes
Titanium Nitride