Learn More
Pain Care Labs Buzzy™ XL Healthcare
SDP
Pain Care Labs Buzzy™ XL Healthcare is proven in clinical trials to decrease the pain of IV insertion, and lets patients know pain relief is a priority.
$156.00 - $159.20
Specifications
Type Medical Device
Material Medical Grade Plastic
Size X-Large
Dimensions (L x W x H) 5.5 x 3 x 8.5 cm
View More Specs
Products 3
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-450-102
SDP
View Documents
Pain Care Labs
BTH 3
LadyBuzz Red
Each for $159.20
Case of 12 EA for $1,898.00
Add to Cart
22-450-101
SDP
View Documents
Pain Care Labs
BTH 2
Plain Black
Each for $156.00
Case of 12 EA for $1,860.00
Add to Cart
22-450-100
SDP
View Documents
Pain Care Labs
BTH 1
Yellow and Black Striped
Each for $159.20
Case of 12 EA for $1,898.00
Add to Cart
Description
Description
  • Latex-free
  • Intended for healthcare and multi-patient use.
  • Includes 4 Universal Healthcare Wings, 4 Silicone Comfort Strap, 2 AAA Batteries (preinstalled in unit), 1 Instruction Manual
  • Reusable medical device that should be thoroughly cleaned and reprocessed following your facility’s infection control protocol for non-critical equipment.
  • Stand up to hospital grade cleansers.
  • Use with disposable hospital tourniquets or press in place for pain relief on contact.
Specifications
Specifications
Medical Device
Medical Grade Plastic
X-Large
5.5 x 3 x 8.5 cm