Learn More
Dino Lite Wireless + USB Digital Microscope WF4915ZT - 1.3MP, 20x - 220x Optical Magnification, Measurement, Polarized Light, AMR, EDOF (WF-20 Included)
Supplier:  Dino Lite WF4915ZT
Encompass_Preferred
The Dino-Lite Edge 1.3MP Bundle WF4915ZT includes the AF4915ZT and WF-20. This all-in-one device is designed to meet most professional applications within 20x- 220x magnifications. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. AMR displays the current magnification on screen with the included software for Windows or Mac OS. Flexible Lighting Control provides lighting adjustment options including intensity settings from 1-6, as well as the ability to turn on or off LED's by quadrant. EDOF is an image processing feature designed to automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field (focus stacking).
Catalog No. NC1660926