Learn More
Sklar Instruments Buck Percussion Hammer, Sklar 6-3/4 in., Stainless Steel / Rubber, EA.
Supplier: Sklar Instruments 063370
Buck Percussion Hammer, Sklar. 6-3/4 in., Stainless Steel / Rubber. Sklar's Buck Percussion Hammer is a diagnostic tool that features a head design similar to that of the Dejerine Hammer; however, the non-latex bumpers on each end of the head are different sizes. The Buck Hammer comes with a pin that screws into the top of the handle which protrudes slightly vertically through the middle point of the bumpers. This hammer also contains a brush that screws into the bottom of the handle. It is used for reflex and general neurological testing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.