Learn More
Medchemexpress LLC HY-13579 5mg Medchemexpress, BTZ043 CAS:1161233-85-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135795MG
Encompass_Preferred
Medchemexpress, HY-13579 5mg BTZ043 CAS:1161233-85-7 BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1401