Learn More
New England Biolabs, Inc. Blunt/TA Ligase Master Mix – 250 reactions
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: New England Biolabs, Inc. M0367L
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Blunt/TA Ligase Master Mix is a ready-to-use solution of T4 DNA Ligase, proprietary ligation enhancer, and optimized reaction buffer. This master mix is specifically formulated to improve ligation and transformation of both blunt-end and single-base overhang substrates. The master mix format simplifies reaction set-up, ensures an optimized ratio of enzyme and buffer components, and yields robust, rapid ligation of all types of DNA ends using a short incubation time at room temperature. No thawing is necessary as it remains liquid during storage at -20C. Ligations for subcloning can be carried out in small volumes with low DNA concentrations, allowing users to conserve precious DNA samples and directly transform many strains of chemically competent E. coli without dilution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.