Learn More
Dojindo Molecular Technologies Inc Biotin-SAM Formation Reagent(1UMOL x3) allows for simple preparation of Biotin-SAM on gold surface and is used for protein immobilization.
Supplier:  Dojindo Molecular Technologies Inc B56410
Encompass
There are three ways to immobilize a protein on a Surface Plasmon Resonance (SPR) or Quartz Crystal Microbalance (QCM) biosensor consisting of Self-Assembled Monolayer (SAM). 1) Formation of covalent bond of amine functions on proteins and activated carboxy-SAM 2) immobilize proteins by His-Tag through Ni-NTA-terminated SAM 3) Immobilization of proteins through Biotin and Avidin interaction. The Biotin-Avidin formation is known as a facile reaction, thus it is widely utilized for immobilization of protein. Dojindo has various reagents for biosensor research. (https://www.dojindo.com/surface-chemistry/)
Catalog No. NC1751686