Learn More
Biopac Systems Inc BIOPAC ELECTRODES BULK 100/PK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Biopac Systems Inc EL503
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
These Pre-Gelled Disposable Electrodes Have A Circular Contact And Are Most Suitable For Short-Term Recordings, Including Surface Emg, Ecg, Eog, Etc. The Small Vinyl Backing Allows Close Electrode Placement Where Necessary, And A Slightly Less Firm Adhesive Allows "Ouchless" Removal. The Electrodes Incorporate Liquid Electrolyte Gel And Moderately-High Chloride Salt Concentration For Quick, Accurate Readings. These Disposable Snap Electrodes Provide The Same Signal Transmission As Reusable Electrodes, With Added Convenience And Hygiene. Each Peel-And-Stick Electrode Is Pre-Gelled And Designed For One Use Only. El503 Electrodes Do Not Contain Any Latex.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.