Learn More
Biomechanical Kits
Offer a complete lab-tested system for teaching and learning the musculoskeletal system
$153.00 - $539.00
Specifications
For Use With (Application) | For teaching and learning the musculoskeletal system |
---|
Includes: All Kits Include:
- Support base and rod
- Support clamps
- Force cord assembly (cable, handle, connectors, fine adjustment, force gauge)
- Instructor's curriculum guide
- Short curriculam for introductoray A&P courses
- Student Laboratory guide
- Biomechanical arm model
- Brass weights (1-50g, 1-100g)
- Biomechanical leg model
- Brass weights (2-200g)
- Contains everything listed above
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S92620C
|
Denoyer Geppert Company
065009 |
Force Cord Assembly | Cable, handle, connectors, fine adjustment, force gauge |
Each for $153.00
|
|
||||
S92620A
|
Denoyer Geppert Company
065011 |
Biomechanical Arm Kit | Biomechanical arm model, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (1 -50g, 1-100 g), instructor's curriculum guide and four student laboratory guides |
Each for $524.00
|
|
||||
S92620B
|
Denoyer Geppert Company
065012 |
Biomechanical Leg Kit | Biomechanical leg model, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (2-200 g), instructor's curriculum guide and four student laboratory guides |
Each for $539.00
|
|
||||
S92620D
|
Denoyer Geppert Company
064900 |
Support Base Unit | Support base and rod, support clamps and guide |
Each for $153.00
|
|
||||
Description
By mastering a limited set of basic concepts, students can predict and explain body movement using the same approaches employed by kinesiologists, exercise physiologists, physical therapists, and orthopedic nurses. Working ideally in teams of four, students practice observation, measurement, data gathering and analysis, prediction, plus calculations and problem solving.
Order Info
Some kit components also sold individually.
Specifications
For teaching and learning the musculoskeletal system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.