Learn More
BD Affirm™ VPIII Microbial Identification Test
DNA probe molecular test for the detection of vaginitis
$53.25 - $27485.00
Specifications
For Use With (Application) | Detection of vaginitis |
---|
Includes: Probe Analysis Cards (PAC) individually wrapped containing five beads (negative control, Trichomonas, Gardnerella, Candida, and positive control); and Reagent Cassettes (RC) containing patient sample reservoir, hybridization solution, wash solution, conjugate, and substrate buffer
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-025-600
|
BD
250100 |
Each | MicroProbe™ Processor |
Each for $27,485.00
|
|
||||
23-025-601
|
BD
446250 |
24 Tests | Affirm™ VPIII Microbial Identification Sample Collection Set |
Each for $263.00
|
|
||||
23-025-602
|
BD
446251 |
100 Pack | Affirm™ VPIII Sample Collection Swab |
Pack of 100 for $53.25
|
|
||||
23-025-603
|
BD
446252 |
24 Tests | Affirm™ VPIII Microbial Identification Test Kit |
Each for $2,796.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Combine with BD Affirm VPIII Ambient Temperature Transport System (Cat. No. 23-025-604)
- For detection and identification of Candida species, Gardnerella vaginalis, and Trichomonas vaginalis nucleic acid in vaginal fluid specimens from vaginitis/vaginosis patients
- Uses two distinct single-stranded nucleic acid probes for each organism: a capture probe and color development probe complementary to unique genetic sequences of target organisms
- Automated processing, with the BD MicroProbe™ Processor (Cat. No. 23-025-600)
- Positive results indicated by visible blue color on target organism bead
- Built-in positive and negative controls on each results card
Specifications
Detection of vaginitis |