Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD AEC Substrate Kit
Substrate for peroxidase and is a widely used chromogen for immunohistochemical staining and immunoblotting
$130.00
Specifications
Chromagen Type | AEC |
---|---|
For Use With (Application) | Immunohistochemical staining and immunoblotting, ELISPOT |
Substrate Type | AEC Substrate |
pH | 7.2 |
Storage Requirements | 4°C |
Alert:
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.
Includes: AEC Buffer and AEC Chromogen
Catalog Number | Mfr. No. | Chromagen Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Chromagen Quantity | Includes | Price | Quantity & Availability | ||||
BDB551015
|
BD
551015 |
1.5mL | AEC buffer and AEC chromogen |
Each for $130.00
|
|
||||
BDB551951
|
BD
551951 |
3mL | 10 plates, AEC buffer and AEC chromogen |
Each for $130.00
|
|
||||
Description
Immunohistochemistry, ELISPOT
Specifications
AEC | |
AEC Substrate | |
4°C |
Immunohistochemical staining and immunoblotting, ELISPOT | |
7.2 |
Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title