Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Columbia CNA Agar with 5% Sheep Blood
Selective and differential media used for the isolation and differentiation of gram-positive microorganisms from clinical and nonclinical materials
$355.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Red |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
L21353
|
BD
221353 |
100/Pk. |
Pack of 100 for $355.00
|
|
|||||
L21352
|
BD
221352 |
20/Pk. |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 12g/L, Peptic digest of Animal Tissue 5g/L, Yeast extract 3.5g/L, Beef extract 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Colistin 10mg/L, Nalidixic Acid 10mg/L, Sheep Blood, defibrinated 5% | |
Plate |
Red | |
Liquid | |
Columbia Colistin-Naladixic Acid Agar with 5% Sheep Blood | |
Gram positive microorganisms |
Safety and Handling
ShelfLife : 84 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title