Learn More
Victory Innovations™ Backpack Electrostatic Sprayer
Backpack Electrostatic Cordless Sprayer is ideal for disinfecting, sanitizing, odor removal, decontamination, and more. Save time and money and get the best chemical coverage for the job.
Supplier: Victory Innovations™ VP300ES
Description
Victory Innovations™ Backpack Electrostatic Cordless Sprayer is ideal for disinfecting, sanitizing, odor removal, decontamination, and more. Save time and money and get the best chemical coverage for the job.
- 3-in-1 nozzle allows flexibility in spraying
- 40-micron, 80-micron, and 110-micron settings to match chemical dwell times
- Covers 23,000 square feet on 40-micron nozzle setting
- Rechargeable lithium-ion battery
- 4 hours of continuous spray time
Specifications
Lithium-ion Battery | |
Disinfecting, odor removal and decontamination | |
3-in-1 Nozzle | |
Handheld Sprayer |